missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cyr61/CCN1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marca: R&D Systems AF6009
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Cyr61/CCN1 Polyclonal specifically detects Cyr61/CCN1 in Human samples. It is validated for Western Blot, Simple Western, ELISA.
Especificaciones
| Cyr61/CCN1 | |
| Polyclonal | |
| Unconjugated | |
| O00622 | |
| CYR61 | |
| Mouse myeloma cell line NS0-derived recombinant human Cyr61/CCN1 Ala22-Asp381 Accession # O00622 | |
| 100 μg | |
| Primary | |
| Detects human Cyr61/CCN1 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| CCN family member 1, CCN1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3491 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido