missing translation for 'onlineSavingsMsg'
Learn More

Human CXCR4 Antibody, R&D Systems™

Código de producto. p-200048264
Change view
Click to view available options
Cantidad:
1 mg
100 μg
25 μg
500 ug
Tamaño de la unidad:
1 miligramo
100 microgramos
25 microgramos
500 microgramos
Product Code. Cantidad unitSize
18189490 1 mg 1 miligramo
18153501 500 ug 500 microgramos
18790047 25 μg 25 microgramos
18782487 100 μg 100 microgramos
4 options
This item is not returnable. View return policy

Product Code. 18189490

Brand: R&D Systems MAB170R01M

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCR4 Monoclonal specifically detects CXCR4 in Human samples. It is validated for Flow Cytometry, Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CXCR4
Aplicaciones Flow Cytometry, Neutralization
Clasificación Monoclonal
Clon 12G5R
Conjugado Unconjugated
Dilución Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.3-1.2 ug/mL
Formulación Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative
N.º de referencia del gen P61073
Alias de gen CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201E, D2S201ESDF-1 receptor, FB22, Fusin, HM89, HM89NPYRL, HSY3RR, LAP3, LAP-3, LCR1, LESTR, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3R, NPY3RWHIM, NPYR, NPYRL, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor, WHIMS
Símbolos de los genes CXCR4
Especie del huésped Mouse
Inmunógeno CP-MAC-infected SUP-T1 human T cell lymphoblastic lymphoma line Accession No. P61073
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 1 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7852
Especificidad de la prueba Detects human CXCR4.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.