missing translation for 'onlineSavingsMsg'
Learn More

Human CXCL5/ENA-78 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18769448
Change view
Click to view available options
Cantidad:
100 μg
25 μg
500 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
500 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18769448 100 μg 100 microgramos
18704438 25 μg 25 microgramos
18724968 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18769448 Proveedor R&D Systems N.º de proveedor MAB254100

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody has been used in 5 publications

CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CXCL5/ENA-78
Aplicaciones Western Blot, Immunohistochemistry, Neutralization
Clasificación Monoclonal
Clon 33160
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 3-15 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P42830
Alias de gen chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, CXCL5, ENA78, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78
Símbolos de los genes CXCL5
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human CXCL5/ENA-78 Ala37-Asn114 (predicted) Accession # P42830
Método de purificación Protein A or G purified from ascites
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6374
Especificidad de la prueba Detects human CXCL5/ENA-78 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CXCL1, 2, 3, 8, 10, or recombinant mouse CXCL2 is observed.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.