missing translation for 'onlineSavingsMsg'
Learn More

Human CX3CL1/Fractalkine Chemokine Domain Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18737968
Change view
Click to view available options
Cantidad:
100 μg
25 μg
500 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
500 microgramos
Código de producto. Cantidad unitSize
18737968 25 μg 25 microgramos
18776469 100 μg 100 microgramos
18762508 500 μg 500 microgramos
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18737968

Marca: R&D Systems MAB3651SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody has been used in 8 publications

CX3CL1/Fractalkine Monoclonal specifically detects CX3CL1/Fractalkine in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CX3CL1/Fractalkine
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Monoclonal
Clon 81513
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P78423
Alias de gen ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1
Símbolos de los genes CX3CL1
Especie del huésped Mouse
Inmunógeno NS0 and E. coli-derived recombinant human CX3CL1/Fractalkine Chemokine Domain Gln25-Arg339 (Ser199Asn) Accession # P78423
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6376
Especificidad de la prueba Detects human CX3CL1/Fractalkine Chemokine Domain in direct ELISAs and Western blots.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG1
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.