missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CTRP5 Synthetic Peptide
Descripción
CTRP5 Synthetic Peptide, PEP-0003, from Invitrogen.
Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. CTRP5 has been suggested to be involved in age-related macular degeneration.
Especificaciones
Especificaciones
| Gene ID (Entrez) | 114902 |
| Especie | Human |
| Método de purificación | purified |
| Número de acceso | Q9BXJ0 |
| Nombre común | Human CTRP5 |
| Concentración | 200 μg/mL |
| Contenido y almacenamiento | -20°C |
| Descripción | Synthetic Peptide |
| Formato | Liquid |
| Formulación | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?