missing translation for 'onlineSavingsMsg'
Learn More

Human CTLA-4 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18735968
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
Código de producto. Cantidad unitSize
18735968 25 μg 25 microgramos
18735079 100 μg 100 microgramos
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18735968

Marca: R&D Systems MAB3252SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Monoclonal Antibody

CTLA-4 Monoclonal specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CTLA-4
Aplicaciones Flow Cytometry, Immunocytochemistry
Clasificación Monoclonal
Clon 2188A
Conjugado Unconjugated
Dilución Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen P16410
Alias de gen CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Especie del huésped Rabbit
Inmunógeno Chinese hamster ovary cell line CHO-derived recombinant human CTLA-4 Ala37-Phe162 Accession # P16410
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 25 μg
Primario o secundario Primary
ID de gen (Entrez) 1493
Especificidad de la prueba Detects human CTLA-4 in direct ELISAs.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Supernatant
Isotype IgG
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.