Learn More
Invitrogen™ Human CREB (aa 123-136) Synthetic Peptide
Código de producto. 11534800
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
11534800 50 μg 50 microgramos
1 options
Código de producto. 11534800 Proveedor Invitrogen™ N.º de proveedor PEP142
Synthetic Peptide
Human CREB (residues 123-136) Synthetic Peptide, PEP-142, from Invitrogen.
CREB (Cyclic AMP response element binding protein) is a 43 kDa basic/leucine zipper transcription factor that binds the cyclic AMP response element (CRE) and activates transcription in response to a variety of extracellular signals including neurotransmitters, hormones, membrane depolarization, and growth and neurotrophic factors. Activation of CREB is dependent upon the phosphorylation of serine 133. Phosphorylation of CREB occurs via p44/42 MAP kinase and p90RSK, and via p38 MAP kinase and MSK1. Although CREB will bind DNA independent of its phosphorylation state, only the phosphorylated form is competent as a transcription factor. CREB binding protein (CBP), a transcriptional coactivator that directly interacts with CREB, binds to CREB in the region of serine 133. CREB is involved in different cellular processes including the synchronization of circadian rhythmicity, differentiation of adipose cells, and learning and memory. In humans, the gene is located on the q arm of chromosome 2. Diseases associated with CREB dysfunction include Alzheimer’s Disease, histiocytoma and soft tissue melanoma. PEP-142 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 123-136 of human CREB . The sequence of this peptide is (amino to carboxy terminus): K (123)-R-R-E-I-L-S-R-R-P-S-Y-R-K (136)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human CREB, catalog &Num; PA1-850. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 mL of distilled water.
TRUSTED_SUSTAINABILITY
Especificaciones
Gene ID (Entrez) 1385
Especie Human
Método de purificación purified
Número de acceso P16220
Nombre común Human CREB (aa 123-136)
Concentración 0.5 mg/mL
Contenido y almacenamiento -20°C. Avoid freeze-thaw cycles.
Descripción Synthetic Peptide
Formato Lyophilized
Formulación peptide with no preservative
Para utilizar con (aplicación) Control
Símbolo de gen CREB1
Liofilizado Yes
Conservante No preservative
Tipo de producto Peptide
Cantidad 50 μg
Estado normativo RUO - research use only
Secuencia K(123)-R-R-E-I-L-S-R-R-P-S-Y-R-K(136)