Learn More
Human CD52 (Research Grade Alemtuzumab Biosimilar) Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18749828
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
18749828 25 μg 25 microgramos
18782228 100 μg 100 microgramos
2 options
Código de producto. 18749828
Marca: R&D Systems MAB9889SP
Human Monoclonal Antibody
CD52 Monoclonal specifically detects CD52 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CD52
Aplicaciones CyTOF, Flow Cytometry
Clasificación Monoclonal
Clon Hu116
Conjugado Unconjugated
Dilución Flow Cytometry 0.25μg/106 cells, CyTOF-ready
Formulación Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Alias de gen Cambridge pathology 1 antigen, CAMPATH-1, CAMPATH-1 antigen, CD52, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, HEL-S-171mP, Human epididymis-specific protein 5
Especie del huésped Human
Inmunógeno Human CD52
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 25 μg
Primario o secundario Primary
ID de gen (Entrez) 1043
Especificidad de la prueba Detects human CD52 based on Alemtuzumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Alemtuzumab. This product is for research use only.
Reconstitución Reconstitute at 0.5mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Supernatant
Isotype IgG2b