missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD52 (Research Grade Alemtuzumab Biosimilar) Alexa Fluor 594-conjugated Antibody, R&D Systems™
Human Monoclonal Antibody
Marca: R&D Systems FAB9889T-100UG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD52 Monoclonal specifically detects CD52 in Human samples. It is validated for Flow Cytometry.
Especificaciones
| CD52 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human CD52 based on Alemtuzumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Alemtuzumab. This product is for research use only. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
| Flow Cytometry | |
| Hu116 | |
| Flow Cytometry 0.25-1μg/106 cells | |
| Cambridge pathology 1 antigen, CAMPATH-1, CAMPATH-1 antigen, CD52, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, HEL-S-171mP, Human epididymis-specific protein 5 | |
| Human CD52 | |
| 100 μg | |
| 1043 | |
| Human | |
| Supernatant |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido