Learn More
Human CD31/PECAM-1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18728589
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18728589 25 Tests 25 tests
18758476 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18728589 Proveedor R&D Systems N.º de proveedor FAB3567A025
Mouse Monoclonal Antibody has been used in 4 publications
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human samples. It is validated for Flow Cytometry, Knockout Validated.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CD31/PECAM-1
Aplicaciones Flow Cytometry, Immunoassay
Clasificación Monoclonal
Clon 9G11
Conjugado APC
Dilución Flow Cytometry 10 μL/10^6 cells, Knockout Validated
Formulación Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Alias de gen adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Símbolos de los genes PECAM1
Especie del huésped Mouse
Inmunógeno Activated HUVEC human umbilical vein endothelial cells
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5175
Especificidad de la prueba Detects human CD31/PECAM-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) E-Selectin, rhICAM-1, -2, -3, rhVCAM-1, or recombinant mouse VCAM-1 is observed.
Especies diana Human
Contenido y almacenamiento Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Formulario Purified
Isotype IgG1