missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD25/IL-2R alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Marca: R&D Systems MAB1020
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.
Especificaciones
| CD25/IL-2R alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 0.5 ug/10^6 cells, CyTOF-ready | |
| P01589 | |
| IL2RA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD25/IL-2 R alpha Glu22-Cys213 Accession # P01589 | |
| 500 μg | |
| Primary | |
| Detects human CD25/IL-2 R alpha in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2 R gamma or rhIL-15 R alpha is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 24212 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3559 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido