Learn More
Invitrogen™ Human CD229 ELISA Kit
Human CD229 quantitates human CD229 in serum, plasma, supernatant.
Marca:  Invitrogen™ EH82RBX10
Detalles adicionales : Peso : 2.50000kg
 Ver más versiones de este producto
Código de producto. 17844170
  • 4540.00€ / paquete de 10
Fecha estimada de envío: 04-05-2023
para ver el stock
Añadir a la cesta
Descripción
Descripción
Human CD229 quantitates human CD229 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD229.
CD229 (Ly9) is a cell surface receptor of the CD150 family, which includes also e.g. CD48 and CD224. Receptors of this family regulate cytokine production and cytotoxicity of lymphocytes and NK cells. High levels of CD229 are found on T and B cells, where its expression increases during their maturation. It is absent on granulocytes, bone marrow-derived dendritic cells, platelets and erythrocytes. CD229 has been also reported on mouse monocytes and NK cells. CD229 interacts homophilically through its N-terminal domain and localizes to the contact site between T cells and antigen presenting B cells during antigen-dependent immune synapse formation.
Especificaciones
Especificaciones
AI893573; CD229; CDABP0070; Cell surface molecule Ly-9; hly9; Lgp100; Ly9; Ly-9; lymphocyte antigen 9; mLY9; RP11-312J18.1; Signaling lymphocytic activation molecule 3; SLAM family member 3; SLAMF3; T100; T-lymphocyte surface antigen Ly-9
80 pg/mL
ELISA
Colorimetric Microplate Reader
Ly9
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
80 to 20,000 pg/mL
Biotin
Supernatant
Q9HBG7
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
10 x 96 Tests
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL
CD229 (Ly9)
4 hr 45 min