missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD117/c-kit Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Marca: R&D Systems AF332-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD117/c-kit Polyclonal specifically detects CD117/c-kit in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.
Especificaciones
| CD117/c-kit | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, KIT, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCF R, SCFR, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3815 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Neutralization 0.06-0.36 ug/mL, CyTOF-ready | |
| P10721 | |
| KIT | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD117/c-kit Gln26-Thr520 Accession # P10721 | |
| 25 μg | |
| Primary | |
| Detects human and mouse CD117/c-kit in direct ELISAs and Western blots. In Western blots, approximately 2% cross-reactivity with recombinant human PDGF sR alpha is observed. | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido