missing translation for 'onlineSavingsMsg'
Learn More

Human CCL18/PARC Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18754488
Change view
Click to view available options
Cantidad:
100 μg
25 μg
500 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
500 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18754488 500 μg 500 microgramos
18738119 25 μg 25 microgramos
18757668 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18754488 Proveedor R&D Systems N.º de proveedor MAB394500

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody has been used in 7 publications

CCL18/PARC Monoclonal specifically detects CCL18/PARC in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CCL18/PARC
Aplicaciones Western Blot, Flow Cytometry, CyTOF, ELISA
Clasificación Monoclonal
Clon 64507
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P55774.1
Alias de gen Alternative macrophage activation-associated CC chemokine 1, AMAC-1, AMAC1CC chemokine ligand 18, AMAC-1Small-inducible cytokine A18, chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated), CKb7, DC-CK1, DC-CK1CC chemokine PARC, DCCK1Macrophage inflammatory protein 4, Dendritic cell chemokine 1, MIP4, MIP-4, MIP-4C-C motif chemokine 18, PARC, PARCPulmonary and activation-regulated chemokine, SCYA18chemokine (C-C), dendritic, small inducible cytokine A18, small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary andactivation-regulated
Símbolos de los genes CCL18
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human CCL18/PARC Ala21-Ala89 Accession # P55774.1
Método de purificación Protein A or G purified from ascites
Cantidad 500 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6362
Especificidad de la prueba Detects human CCL18/PARC in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant human CCL3, 4, 14, 15, 23, recombinant mouse (rm) CCL3, rmCCL4, recombinant viral (rv) MIP-I or rvMIP-II.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Ascites
Isotype IgG1
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.