missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cannabinoid R2/CB2/CNR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB36551R-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Cannabinoid R2/CB2/CNR2 Monoclonal specifically detects Cannabinoid R2/CB2/CNR2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Especificaciones
| Cannabinoid R2/CB2/CNR2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| cannabinoid receptor 2 (macrophage), CannabinoidR2, CB-2, CB2cannabinoid receptor 2, CB2R, CNR2, CX5, CX5testis-dominant CNR2 isoform CB2, hCB2 | |
| Human embryonic kidney cell line HEK293-derived transfected with human Cannabinoid R2/CB2/CNR2, Met1-Cys360, Accession # NP_001832 | |
| 100 μg | |
| 1269 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 352110R | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| NP_001832 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Cannabinoid R2/CB2/CNR2 in HEK293 human cell line transfected with human Cannabinoid R2/CB2/CNR2 and not Cannabinoid R1/CB1/CNR1 transfectants in flow cytometry. | |
| Human | |
| Lyophilized |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido