Learn More
Human C-Reactive Protein/CRP Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18762808
Change view
Click to view available options
Cantidad:
100 μg
25 μg
500 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
500 microgramos
18762808 100 μg 100 microgramos
18761249 25 μg 25 microgramos
18755499 500 μg 500 microgramos
3 options
Código de producto. 18762808
Marca: R&D Systems MAB17071100
Mouse Monoclonal Antibody
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno C-Reactive Protein/CRP
Aplicaciones ELISA
Clasificación Monoclonal
Clon 232007
Concentración LYOPH
Conjugado Unconjugated
Dilución ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P02741
Alias de gen C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895
Símbolos de los genes CRP
Especie del huésped Mouse
Inmunógeno Human plasma-derived CRP Accession # P02741
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1401
Especificidad de la prueba Detects human CRP in direct ELISAs.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Supernatant
Isotype IgG2b