missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human beta-TrCP1/BTRC Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB7675
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
beta-TrCP1/BTRC Monoclonal specifically detects beta-TrCP1/BTRC in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Especificaciones
| beta-TrCP1/BTRC | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q9Y297-2 | |
| BTRC | |
| E. coli-derived recombinant human beta-TrCP1 isoform 2 Met1-Leu120 Accession # Q9Y297-2 | |
| 100 μg | |
| Primary | |
| Detects human BetaTrCP1 isoform 2 in direct ELISAs and Western blots. Detects mouse BetaTrCP1 in Western Blots. Detection of mouse BetaTrCP1 in Immuncytochemistry has not been tested. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 763524 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-transducin repeat containing, betaTrCP, BETA-TRCP, betaTrCP1, beta-TrCP1, BTRC, bTrCP, bTrCP1, E3RSIkappaB, F-box and WD repeats protein beta-TrCP, F-box/WD repeat-containing protein 1A, FBW1A, FBXW1, FBXW1A, FBXW1ABTRCP, Fwd1, MGC4643, pIkappaBalpha-E3 receptor subunit | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8945 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido