missing translation for 'onlineSavingsMsg'
Learn More

Human BAK Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18736898
Change view
Click to view available options
Cantidad:
25 μg
Tamaño de la unidad:
25 microgramos
Código de producto. Cantidad unitSize
18736898 25 μg 25 microgramos
1 options
Este artículo no se puede devolver. View return policy

Product Code. 18736898

Brand: R&D Systems AF816SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 3 publications

BAK Polyclonal specifically detects BAK in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno BAK
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 1-4 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen Q16611
Alias de gen Apoptosis regulator BAK, BAK1, BAK-LIKE, BAKMGC3887, BCL2-antagonist/killer 1, Bcl2-L-7, BCL2L7, BCL2L7bcl-2 homologous antagonist/killer, BCL2-like 7 protein, Bcl-2-like protein 7, CDN1, MGC117255, pro-apoptotic protein BAK
Símbolos de los genes BAK1
Especie del huésped Rabbit
Inmunógeno KLH-coupled human BAK synthetic peptide AAPADPEMVTLPLQPSSTMGC Accession # Q16611
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 578
Especificidad de la prueba Detects human BAK in Western blots. This antibody does not detect mouse BAK.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.