missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BAFF/BLyS/TNFSF13B Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Marca: R&D Systems MAB124-500
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BAFF/BLyS/TNFSF13B Monoclonal specifically detects BAFF/BLyS/TNFSF13B in Human samples. It is validated for Neutralization.
Especificaciones
| BAFF/BLyS/TNFSF13B | |
| Monoclonal | |
| LYOPH | |
| Neutralization 20 ug/mL | |
| Q9Y275 | |
| TNFSF13B | |
| Mouse myeloma cell line NS0-derived recombinant human BAFF Ala134-Leu285 Accession # Q9Y275 | |
| 500 μg | |
| Primary | |
| Detects human BAFF in direct ELISAs. | |
| Human | |
| Purified |
| Neutralization | |
| 148725 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL1, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF13B, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10673 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido