missing translation for 'onlineSavingsMsg'
Learn More

Human BAFF/BLyS/TNFSF13B Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18777047
Change view
Click to view available options
Cantidad:
25 μg
Tamaño de la unidad:
25 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18777047 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18777047 Proveedor R&D Systems N.º de proveedor AF124SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody has been used in 11 publications

BAFF/BLyS/TNFSF13B Polyclonal specifically detects BAFF/BLyS/TNFSF13B in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno BAFF/BLyS/TNFSF13B
Aplicaciones Western Blot, Immunohistochemistry, Neutralization
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 3-12 ng/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen Q9Y275
Alias de gen ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL1, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF13B, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4
Símbolos de los genes TNFSF13B
Especie del huésped Goat
Inmunógeno Mouse myeloma cell line NS0-derived recombinant human BAFF/BLyS/TNFSF13B Ala134-Leu285 Accession # Q9Y275
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10673
Especificidad de la prueba Detects human BAFF/BLyS/TNFSF13B in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant murine BAFF and less than 1% cross-reactivity with recombinant human (rh) APRIL is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.