missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ARNT/HIF-1 beta Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB5630
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ARNT/HIF-1 beta Monoclonal specifically detects ARNT/HIF-1 beta in Human samples. It is validated for Western Blot, Immunocytochemistry.
Especificaciones
| ARNT/HIF-1 beta | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P27540 | |
| ARNT | |
| E. coli-derived recombinant human ARNT/HIF-1 beta Asp517-Ala691 Accession # P27540 | |
| 100 μg | |
| Primary | |
| Detects human ARNT/HIF-1 beta in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 775146 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARNT, ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, Class E Basic Helix-Loop-Helix Protein 2, Dioxin Receptor, Nuclear Translocator, HIF1 beta, HIF-1 Beta, HIF1B, HIF-1beta, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, TANGO | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 405 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido