Learn More
Human Apolipoprotein B/ApoB Antibody, R&D Systems™
Código de producto. 18754367
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18754367 25 μg 25 microgramos
18734417 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18754367 Proveedor R&D Systems N.º de proveedor MAB41241SP
Mouse Monoclonal Antibody
Apolipoprotein B/ApoB Monoclonal antibody specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Apolipoprotein B/ApoB
Aplicaciones Immunohistochemistry, Immunocytochemistry
Clasificación Monoclonal
Clon 989529
Conjugado Unconjugated
Dilución Immunohistochemistry 5-25 μg;/mL, Immunocytochemistry 8-25 μg;/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen NP_000375
Alias de gen Apo B-100, APOB, apoB-100, ApoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human Apolipoprotein B/ApoB, Met1206-Asp1413, Accession No. NP_000375
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Primario o secundario Primary
ID de gen (Entrez) 338.0
Especificidad de la prueba Detects human Apolipoprotein B/ApoB in direct ELISAs.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Formulario Supernatant
Isotype IgG2b