missing translation for 'onlineSavingsMsg'
Learn More

Human Apolipoprotein A-IV/ApoA4 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18770297
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
Código de producto. Cantidad unitSize
18770297 100 μg 100 microgramos
18740027 25 μg 25 microgramos
2 options
This item is not returnable. View return policy

Product Code. 18770297

Brand: R&D Systems MAB8125100

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Apolipoprotein A-IV/ApoA4 Monoclonal specifically detects Apolipoprotein A-IV/ApoA4 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Apolipoprotein A-IV/ApoA4
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Monoclonal
Clon 988829
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen P06727
Alias de gen apo-AIV, ApoA-IV, Apolipoprotein A4, apolipoprotein A-IV, Apolipoprotein AIV, MGC142154, MGC142156
Especie del huésped Mouse
Inmunógeno E. coli-derived human Apolipoprotein A-IV/ApoA4 protein, Glu21-Ser396, Accession No. P06727
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Primario o secundario Primary
ID de gen (Entrez) 337.0
Especificidad de la prueba Detects human Apolipoprotein A-IV/ApoA4 in direct ELISAs.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Formulario Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.