missing translation for 'onlineSavingsMsg'
Learn More

Human Angiopoietin-like Protein 4/ANGPTL4 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18716105
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18716105 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18716105 Proveedor R&D Systems N.º de proveedor FAB11305G100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Angiopoietin-like Protein 4/ANGPTL4
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 944740
Conjugado Alexa Fluor 488
Formulación Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
N.º de referencia del gen Q9BY76
Alias de gen angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein
Especie del huésped Mouse
Inmunógeno Mouse myeloma cell line, NS0-derived human Angiopoietin-like Protein 4/ANGPTL4 protein, Gly26-Ser406, Accession # Q9BY76
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 51129
Especies diana Human
Contenido y almacenamiento Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.