missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ALK-7 APC-conjugated Antibody, R&D Systems™

Description
ALK-7/Activin Receptor Type 1C Monoclonal specifically detects ALK-7/Activin Receptor Type 1C in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antígeno | ALK-7/Activin Receptor Type 1C |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 810506 |
| Conjugado | APC |
| Dilución | Flow Cytometry 10 uL/10^6 cells |
| Formulación | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| N.º de referencia del gen | Q8NER5 |
| Alias de gen | activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, Activin RIC, ACTR-IC, ACVR1C, ACVRLK7, ALK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30 |
| Símbolos de los genes | ACVR1C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?