Learn More
Invitrogen™ Human ADAMTS-13 ELISA Kit
Human ADAMTS-13 quantitates human ADAMTS-13 in serum, plasma, supernatant.
Marca:  Invitrogen™ EH12RBX10
Detalles adicionales : Peso : 2.50000kg
 Ver más versiones de este producto
Código de producto. 17804070
  • 4540.00€ / paquete de 10
Fecha estimada de envío: 28-04-2023
para ver el stock
Añadir a la cesta
Descripción
Descripción
Human ADAMTS-13 quantitates human ADAMTS-13 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ADAMTS-13.
ADAMTS13 is a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme is the von Willebrand Factor (vWF)-cleaving protease, which is responsible for cleaving at the site of Tyr842-Met843 of the vWF molecule. A deficiency of this enzyme is associated with thrombotic thrombocytopenic purpura.
Especificaciones
Especificaciones
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 13; A disintegrin and metalloproteinase with thrombospondin motifs 13-like; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 13; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 13; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 13; ADAM metallopeptidase with thrombospondin type 1 motif, 13; ADAMs; ADAM-TS 13; Adamts13; ADAM-TS13; ADAMTS-13; C9orf8; Gm710; LOC102554393; metalloendopeptidases; UNQ6102/PRO20085; von Willebrand factor-cleaving protease; vWF-cleaving protease; VWFCP; vWF-CP; vWF-CP mRNA for von Willebrand factor-cleaving
7.5 ng/mL
ELISA
Colorimetric Microplate Reader
ADAMTS13
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
7.5 to 1800 ng/mL
Biotin
Supernatant
Q76LX8
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
10 x 96 Tests
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL
ADAMTS13
4 hr 45 min