missing translation for 'onlineSavingsMsg'
Learn More

Human ACE/CD143 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18038321
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
18038321 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18038321

Marca: R&D Systems AF929

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody has been used in 3 publications

ACE/CD143 Polyclonal specifically detects ACE/CD143 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno ACE/CD143
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready
N.º de referencia del gen P12821
Alias de gen ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA
Símbolos de los genes ACE
Especie del huésped Goat
Inmunógeno Mouse myeloma cell line NS0-derived recombinant human ACE/CD143 Leu30-Leu1261 Accession # P12821
Método de purificación Affinity Purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1636
Especificidad de la prueba Detects ACE/CD143 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse ACE is observed and less than 5% cross-reactivity with recombinant human ACE-2 is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.