missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ACE-2 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Descripción
ACE-2 Monoclonal specifically detects ACE-2 in Human samples. It is validated for Flow Cytometry.
Especificaciones
Especificaciones
| Antígeno | ACE-2 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 379131 |
| Conjugado | Alexa Fluor 350 |
| Dilución | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulación | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| N.º de referencia del gen | Q9BYF1 |
| Alias de gen | ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 |
| Especie del huésped | Rat |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?