missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTLV-2 env 801 Monoclonal Antibody (8.A.033)

Descripción
MA1-91454 detects HTLV2 env 801 from human samples.
The HTLV-2 env 801 gene encodes the envelope glycoprotein, which is pivotal for virus entry into host cells. Human T-cell leukemia virus type 2 (HTLV-2) shares many structural features with HTLV-1, with the envelope (Env) protein being a critical component in mediating viral entry via interaction with cell surface receptors. The env gene product is synthesized as a precursor that is cleaved into the surface (SU) and transmembrane (TM) subunits. The SU subunit facilitates receptor binding, while the TM subunit aids in membrane fusion, enabling the virus to enter the host cell. HTLV-2 predominantly targets and transforms CD8+ T-cells, distinct from HTLV-1, which primarily transforms CD4 + T-cells. Understanding the structural and functional nuances of the HTLV-2 envelope protein is crucial as it presents a target for therapeutic interventions aimed at preventing viral entry and subsequent infection.
Especificaciones
Especificaciones
| Antígeno | HTLV-2 env 801 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 8.A.033 |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 0.1M tris HCl with no preservative; pH 7.5 |
| Alias de gen | HTLV; HTLV2; HTLV-II |
| Especie del huésped | Mouse |
| Inmunógeno | Highly purified HTLV-II envelope 801 antigen. |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?