missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSP70 Recombinant Rabbit Monoclonal Antibody (HL1580)

Product Code. 18055551
Change view
Click to view available options
Cantidad:
100 μL
Unit Size:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18055551 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18055551 Supplier Invitrogen™ Supplier No. MA550321

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

HSP70 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

HSP70 is a Heat shock protein (HSP) which are expressed in response to various biological stresses, including high temperatures. There are several major families of HSPs that include HSP70, there are HSP90 and HSP100. The HSP70 family is a set of highly conserved proteins that are induced by a variety of biological stresses, including heat stress, in every organism in which the proteins have been examined. The human HSP70 family members include: HSP70, a protein which is strongly inducible in all organisms but which is also constitutively expressed in primate cells; HSP72, a 72 kDa protein that is induced exclusively under stress conditions; HSC70, or cognate protein, is a 72 kDa, constitutively expressed, protein which is involved in the uncoating of clathrin coated vesicles; GRP78, or BiP, is a glucose regulated 78 kDa protein localized in the endoplasmic reticulum; and p75, or HSP75, a 75 kDa protein that is found within the mitochondria. Further, HSP70 is encoded by an intronless gene and, in conjunction with other heat shock proteins, HSP70 stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and organelles. HSP70 is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The HSP70 gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno HSP70
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon HL1580
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica HSPA1B
N.º de referencia del gen O88600, P0DMV8, P0DMV9, P0DMW0, P0DMW1, P14659, P16627, P17156, P17879, P34931, P34932, P54652, P55063, Q2TFN9, Q61316, Q61696, Q7YQC6
Alias de gen 68 kDa heat shock protein; 70 kda heat shock protein-2; 70 kilodalton heat shock protein; 87A; 87A locus; 87C locus; Bb; CG18743; CG31359; CG31359-PA; CG31366; CG31366-PA; CG31449; CG6489; chaperone; dHSP70; dhsp70Aa; Dmel\CG31359; Dmel\CG31366; Dmel_CG31359; Dmel_CG31366; Dm-hsp70; DmHSP70AA; DMHSP7D1; dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; Hcp70.1; heat shock 10kDa protein 1-like; heat shock 70; heat shock 70 kD protein 1; heat shock 70 kD protein 2; heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 1-Hom; Heat shock 70 kDa protein 1L; heat shock 70 kDa protein 1-like; heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70 kDa-like protein 1; heat shock 70kD protein 1A; heat shock 70kD protein 1B; heat shock 70kD protein 1B (mapped); heat shock 70kD protein-like 1; heat shock 70kDa protein 1A; heat shock 70kDa protein 1B; heat shock 70kDa protein 1-like; heat shock cognate 70-kd protein, tandem duplicate 2; Heat shock protein; heat shock protein 1A; heat shock protein 1B; heat shock protein 1-like; heat shock protein 70; heat shock protein 70.1; heat shock protein 70.2; heat shock protein 70-1; Heat shock protein 70-1l; Heat shock protein 70Aa; Heat shock protein 70Ab; Heat shock protein 70Ba; Heat shock protein 70Bb; Heat shock protein 70Bc; heat shock protein cognate 70, testis; heat shock protein family A (Hsp70) member 1 like; heat shock protein family A (Hsp70) member 1A; heat shock protein family A (Hsp70) member 1B; heat shock protein, 70 kDa 1; heat shock protein, 70 kDa 3; heat shock protein-70; heat shock-induced protein; heat shock-related protein hsc70t; heat-shock 70-kilodalton protein 1A; heat-shock 70-kilodalton protein 1B; heat-shock protein 70; Heat-shock-protein-70Aa; Heat-shock-protein-70Bb; HEL S 103; HEL-S-103; hs70; Hsc70t; HSP; Hsp 70; hsp68; Hsp70; Hsp-70; HSP70 1; HSP70 1B; HSP70 2; hsp70 87A; hsp70 87C; hsp70 Aa; HSP70 AceyltK77; hsp70(87A); Hsp70(87C); HSP70.1; HSP70.1/2; HSP70.1/HSP70.2; HSP70.2; Hsp70.3; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; HSP70-1B; HSP70-1L; HSP70-2; Hsp70-3; HSP70-87A7; HSP70-87C1; hsp70a; hsp70A1; Hsp70A7d; Hsp70Aa; Hsp-70Aa; Hsp70Aa-PA; Hsp70Ab; Hsp70B; Hsp70Ba; Hsp70Bb; Hsp70Bbc; Hsp70Bb-PA; Hsp70Bc; HSP70-HOM; HSP70I; HSP70T; HSP72; Hspa1; Hspa1a; HSPA1B; Hspa1l; Hspa2; HSPA4; Hspa70; HSX70; hum70t; inducible heat shock protein 70; LP05203p; LP05233p; LP08776p; Major heat shock 70 kDa protein Aa; Major heat shock 70 kDa protein Ab; Major heat shock 70 kDa protein Ba; Major heat shock 70 kDa protein Bb; Major heat shock 70 kDa protein Bc; Msh5; protein 70; si:ch73-46h18.6; si:dkeyp-69e1.1; Spermatid-specific heat shock protein 70; zgc:174006
Símbolos de los genes hsp70, HSPA1A, HSPA1B, Hspa1l, HSPA2, HSPA4
Especie del huésped Rabbit
Inmunógeno Recombinant protein encompassing a sequence within the center region of human Hsp70. The exact sequence is proprietary.
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 101088003, 101097784, 15482, 15511, 15512, 15525, 193740, 24472, 24963, 266759, 294254, 3303, 3304, 3305, 3306, 3308, 403612, 474680, 480355, 60460
Especies diana Human, Mouse, Rat, Canine, Feline
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.