Learn More
HSP70/HSC70 Monoclonal Antibody (5G1-95), APC, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA545134
Descripción
1 μg/mL of MA5-45134 was sufficient for detection of HSP70/HSC70 in 20 μg of Phaseolus aureus (mung bean) lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.∣Detects approximately 70kDa. Recognizes constitutive and inducible plants HSP70 (HSC70/HSP72). Does not cross-react with Human, Rat, bacteria (DNAK) or Human Bip.
HSPA4 (Heat Shock Protein Family A (Hsp70) Member 4) is a Protein Coding gene. Diseases associated with HSPA4 include Chagas Disease and Babesiosis. Among its related pathways are Regulation of activated PAK-2p34 by proteasome mediated degradation and Cellular responses to stimuli. An important paralog of this gene is HSPH1. HSPA8 (Heat Shock Protein Family A (Hsp70) Member 8) is a Protein Coding gene. Diseases associated with HSPA8 include Auditory System Disease and Borna Disease. Among its related pathways are Processing of Capped Intron-Containing Pre-mRNA and Selective autophagy. Gene Ontology (GO) annotations related to this gene include RNA binding and ubiquitin protein ligase binding. An important paralog of this gene is HSPA2.
Especificaciones
| HSP70/HSC70 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.15mM MES, 47.35mM phosphate with 1.033mM EDTA and no preservative; pH 7.2 | |
| Mouse | |
| Protein G | |
| RUO | |
| Plant | |
| Liquid |
| ELISA, Western Blot | |
| 5G1-95 | |
| APC | |
| HSPA4/HSPA8 | |
| Purified HSP70 from Phaseolus aureus (mung bean) | |
| 100 μg | |
| Primary | |
| 4° C | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.