missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSP20 Monoclonal Antibody (C9)
Descripción
HSP20 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Heat Shock Protein 20 (HSP20), encoded by the HSPB6 gene, is a member of the small heat shock protein (sHSP) family, prominently involved in cellular stress response and maintenance of cytoskeletal integrity. HSP20 is ubiquitously expressed across various tissues, including cardiac and smooth muscle, where its roles are vital in modulating stress resilience and muscle function. It exerts protective effects against apoptosis and oxidative stress by interacting with cell signaling pathways that enhance cellular survival and protein stabilization. In cardiac muscle, HSP20 is known for its cardioprotective actions during ischemic injury, helping preserve myocardial function by inhibiting hypertrophic signaling pathways and promoting muscle relaxation. Studies highlight its involvement in vasodilation processes, as HSP20 interacts with myosin to regulate smooth muscle contraction and dilatation. Beyond its role in muscle physiology, HSP20 has been investigated for its potential therapeutic applications in cardiovascular diseases and metabolic disorders, where its expression and function could be pivotal in disease modulation and treatment strategies.
Especificaciones
Especificaciones
| Antígeno | HSP20 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | C9 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| génica | HSPB6 |
| N.º de referencia del gen | P97541 |
| Alias de gen | AA387366; cardiac heat shock protein p20; cardiac p20; epididymis luminal protein 55; Gm479; Heat shock 20 kDa-like protein p20; heat shock 20-kDa protein; Heat shock protein; heat shock protein 20kDa; heat shock protein beta-6; heat shock protein family B (small) member 6; heat shock protein family B (small) member B6; heat shock protein, alpha-crystallin-related, B6; HEL55; HSP; HSP20; HSPB6; p20; PPP1R91; protein phosphatase 1, regulatory subunit 91 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?