missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22883-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is specific to human HSF4.
Heat-shock transcription factors (HSFs) activate heat-shock response genes under conditions of heat or other stresses. HSF4 lacks the carboxyl-terminal hydrophobic repeat which is shared among all vertebrate HSFs and has been suggested to be involved in the negative regulation of DNA binding activity. Two alternatively spliced transcripts encoding distinct isoforms and possessing different transcriptional activity have been described.Especificaciones
| HSF4 | |
| Polyclonal | |
| Unconjugated | |
| HSF4 | |
| CTM, Heat shock factor protein 4, hHSF4, HSF 4, HSF4, HSTF 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26386, 291960, 3299 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9R0L1, Q9ULV5 | |
| Hsf4 | |
| HSF4 Fusion Protein Ag18935 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido