missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF2 Monoclonal Antibody (OTI2E2), TrueMAB™, OriGene
Descripción
All organisms respond to elevated temperatures and a variety of environmental stresses by rapid synthesis of heat shock RNAs and proteins. The regulation of heat shock gene transcription is mediated by the transcriptional activator, heat shock factor (HSF), which binds to heat shock response elements (HSEs). Heat shock factor protein 2 (HSF-2) is a DNA-binding protein that specifically binds heat shock promoter elements (HSE) and activates transcription. In higher eukaryotes, HSF is unable to bind to theHSE unless the cells are heat shocked.
Especificaciones
Especificaciones
| Antígeno | HSF2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI2E2 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | HSF2 |
| N.º de referencia del gen | Q03933 |
| Alias de gen | HSF 2, HSTF 2 |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?