missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRK Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA520235
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Despite its predicted molecular weight, Hrk often migrates at 12-15kDa in SDS-PAGE. A suggested positive control is mouse pancreas tissue lysate. PA5-20235 can be used with blocking peptide PEP-0369.
Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Hrk, a pro-apoptotic member of the Bcl-2 homology domain-3 (BH3)-only group of the Bcl-2 family of proteins, was also identified as novel protein induced during programmed neuronal death. It lacks significant homology to other Bcl-2 family members except for an 8-amino acid region that is similar to the BH3 motif of Bik. Hrk regulates apoptosis through interaction with the anti-apoptotic proteins Bcl-2 and Bcl-XL via this domain. It does not interact with the pro-apoptotic proteins Bax, Bak, or Bcl-XS. Hrk localizes to mitochondrial membranes in a pattern similar to that previously reported for Bcl-2 and Bcl-XL.
Especificaciones
| HRK | |
| Polyclonal | |
| Unconjugated | |
| HRK | |
| Activator of apoptosis harakiri; activator of apoptosis Hrk; AI838259; BCL2-interacting protein; BH3 interacting (with BCL2 family) domain, apoptosis agonist; BH3-interacting domain-containing protein 3; Bid3; death protein 5; DP5; harakiri; harakiri, BCL2 interacting protein; harakiri, BCL2 interacting protein (contains only BH3 domain); Hrk; Neuronal death protein DP5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12123, 8739 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O00198, P62816 | |
| HRK | |
| A 15 amino acid peptide from near the center of human Hrk. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido