missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRK Polyclonal Antibody

Descripción
Sequence homology: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 83%; Rabbit: 100%; Rat: 83%.
Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Hrk, a pro-apoptotic member of the Bcl-2 homology domain-3 (BH3)-only group of the Bcl-2 family of proteins, was also identified as novel protein induced during programmed neuronal death. It lacks significant homology to other Bcl-2 family members except for an 8-amino acid region that is similar to the BH3 motif of Bik. Hrk regulates apoptosis through interaction with the anti-apoptotic proteins Bcl-2 and Bcl-XL via this domain. It does not interact with the pro-apoptotic proteins Bax, Bak, or Bcl-XS. Hrk localizes to mitochondrial membranes in a pattern similar to that previously reported for Bcl-2 and Bcl-XL.
Especificaciones
Especificaciones
| Antígeno | HRK |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| génica | HRK |
| N.º de referencia del gen | O00198 |
| Alias de gen | Activator of apoptosis harakiri; activator of apoptosis Hrk; AI838259; BCL2-interacting protein; BH3 interacting (with BCL2 family) domain, apoptosis agonist; BH3-interacting domain-containing protein 3; Bid3; death protein 5; DP5; harakiri; harakiri, BCL2 interacting protein; harakiri, BCL2 interacting protein (contains only BH3 domain); Hrk; Neuronal death protein DP5 |
| Símbolos de los genes | HRK |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?