missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA124511
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PA1-24511 detects Histamine H2 Receptor in Human samples.
H2 Histamine Receptor stimulates adenylate cyclase activity and leads to the release of acids in the stomach. It is also involved in GI motility and intestinal secretion. Treatments for acid-peptic disorders are based on the inhibition of histamine H2 receptor activity. The 3-untranslated region may be involved in differential expression. Histamine H2 receptor has been reported to be expressed in bone marrow, brain, heart, lymph node, nose, placenta, spinal cord, and stomach. An EST has been isolated from a kidney library.
Especificaciones
| HRH2 | |
| Polyclonal | |
| Unconjugated | |
| HRH2 | |
| gastric receptor 1; gastric receptor I; H2R; HH2R; histamine H2 receptor; histamine receptor; histamine receptor H 2; histamine receptor H2; HRH2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15466, 25461, 3274 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P25021, P25102, P97292 | |
| HRH2 | |
| Synthetic 16 amino acid peptide from 3rd extracellular domain of human HRH2/Histamine H2 Receptor. | |
| 25 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido