missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRG Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26252-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HRG contains two cystatin-like domains and is located in plasma and platelets. The physiological function has not been determined but it is known that the protein binds heme, dyes and divalent metal ions. It can inhibit rosette formation and interacts with heparin, thrombospondin and plasminogen. Two of the protein′s effects, the inhibition of fibrinolysis and the reduction of inhibition of coagulation, indicate a potential prothrombotic effect. Mutations in this gene lead to thrombophilia due to abnormal histidine-rich glycoprotein levels.Especificaciones
| HRG | |
| Polyclonal | |
| Unconjugated | |
| HRG | |
| histidine rich glycoprotein, HPRG, HRG, HRGP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3273 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P04196 | |
| HRG | |
| HRG Fusion Protein Ag16278 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido