missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV18 E7 Monoclonal Antibody (OTI10D7), TrueMAB™, OriGene
Description
HPV18 E7 Monoclonal antibody specifically detects HPV18 E7 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | HPV18 E7 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI10D7 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | ZBTB17 |
| N.º de referencia del gen | Q13105 |
| Alias de gen | HPV E7; HPV18; HPV-18; human papillomavirus |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?