missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HPS1 Monoclonal Antibody (5G12G2)

Descripción
MA5-15381 targets HPS1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15381 immunogen is purified recombinant fragment of HPS1 expressed in E. Coli..
Hermansky-Pudlak syndrome (HPS) is a rare, genetically heterogeneous, autosomal recessive disorder. It is characterized by oculocutaneous albinism, lysosomal storage defects and prolonged bleeding due to platelet storage pool deficiency. There are 10 HPS genes encoding HPS proteins that all interact within three distinct ubiquitously expressed protein complexes or biogenesis of lysosome-related organelle complexes. Defects in these genes cause HPS. HSP-1 is a component of multiple cytoplasmic organelles and is important for their normal development and function. It plays a role in intracellular protein sorting.
Especificaciones
Especificaciones
| Antígeno | HPS1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 5G12G2 |
| Concentración | Conc. Not Determined |
| Conjugado | Unconjugated |
| Formulación | Ascites with 0.03% sodium azide |
| génica | HPS1 |
| N.º de referencia del gen | Q92902 |
| Alias de gen | 6030422N11Rik; BB405864; BLOC3S1; ep; Gm21361; Hermansky-Pudlak syndrome 1 homolog (human); Hermansky-Pudlak syndrome 1 protein; Hermansky-Pudlak syndrome 1 protein homolog; Hermansky-Pudlak syndrome protein homolog; HPS; HPS1; HPS1, biogenesis of lysosomal organelles complex 3 subunit 1; MGC5277; pale ear |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?