missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD9 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene
Description
HOXD9 Monoclonal antibody specifically detects HOXD9 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | HOXD9 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI2B1 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | HOXD9 |
| N.º de referencia del gen | P28356 |
| Alias de gen | homeo box D9; homeobox D9; homeobox Hox-D9; homeobox protein Hox-4.4; Homeobox protein Hox-4C; homeobox protein Hox-5.2; Homeobox protein Hox-D9; HOX4; Hox-4.3; Hox-4.3, mouse, homolog of; Hox-4.4; HOX4C; Hox-5.2; HOXD9; Hoxd-9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?