Learn More
HOXD9 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene
Código de producto. 17542114
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Tamaño de la unidad:
100 microlitros
30 microlitros
17542114 100 μL 100 microlitros
17562114 30 μL 30 microlitros
2 options
Código de producto. 17542114
Marca: Origene Technologies TA809665
Este artículo requiere una actualización de precios antes de ser solicitado.
Solicite el precio de este artículo aquí.
Mouse Monoclonal Antibody
HOXD9 is the sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that removed the entire HOXD gene cluster or 5′ end of this cluster have been associated with severe limb and genital abnormalities. The exact role of this gene has not been determined.
Especificaciones
Antígeno HOXD9
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI2B1
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica HOXD9
N.º de referencia del gen P28356
Alias de gen HOX4, HOX4C, Hox-4.3, Hox-5.2
Símbolos de los genes HOXD9
Especie del huésped Mouse
Inmunógeno Human recombit protein fragment corresponding to amino acids 109-352 of human HOXD9 produced in E.coli.
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3235
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1