Learn More
Invitrogen™ HOXD8 Polyclonal Antibody
GREENER_CHOICE
Código de producto. 16653885
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
16653885 100 μL 100 microlitros
1 options
Código de producto. 16653885
Marca: Invitrogen™ PA599639
Rabbit Polyclonal Antibody
Antibody detects endogenous levels of total HoxD8.
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located in a cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. In addition to effects during embryogenesis, this particular gene may also play a role in adult urogenital tract function.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno HOXD8
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica HOXD8
N.º de referencia del gen P13378
Alias de gen 4921540P06Rik; AI047735; homeo box 4E; homeo box D8; homeobox D8; homeobox protein 5.4; homeobox protein Hox-4.3; Homeobox protein Hox-4E; homeobox protein Hox-5.4; Homeobox protein Hox-D8; homeobox protein Hox-D8-like; HOX4; Hox-4.3; Hox-4.5; HOX4E; HOX5.4; Hox-5.4; HOXD8; Hoxd-8; LOC100912936
Símbolos de los genes HOXD8
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human HOXD8(Accession P13378), corresponding to amino acid residues L53-A103.
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3234
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG