missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXD10 Polyclonal Antibody

Descripción
Antibody detects endogenous levels of total HoxD10.
This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox D genes located on chromosome 2. The encoded nuclear protein functions as a sequence-specific transcription factor that is expressed in the developing limb buds and is involved in differentiation and limb development. Mutations in this gene have been associated with Wilm's tumor and congenital vertical talus and/or a foot deformity resembling that seen in Charcot-Marie-Tooth disease.
Especificaciones
Especificaciones
| Antígeno | HOXD10 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | HOXD10 |
| N.º de referencia del gen | P28358 |
| Alias de gen | AI385591; AI874987; d10; homeo box 4D; homeo box D10; homeo box D10a; homeobox D10; homeobox D10a; homeobox gene D-10; homeobox protein Hox-4.5; Homeobox protein Hox-4D; homeobox protein Hox-4E; Homeobox protein Hox-5.3; homeobox protein Hox-C10; Homeobox protein Hox-D10; homeobox protein Hox-D10a; homeobox protein Hox-D11; homeobox protein Hox-D10; HOX4; Hox-4.4; Hox-4.5; Hox-45; HOX4D; HOX4E; Hox-5.3; hoxc10; hox-C10; Hoxc10 protein; Hoxd10; HOX-D10; hoxd-10; hoxd10a; hoxd10a protein; z-82 |
| Símbolos de los genes | HOXD10 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?