missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXC9 Polyclonal Antibody
Descripción
Immunogen sequence: PSQSSVVYHPY GPQPHLGADT RYMRTWLEPL SGAVSFPSFP AGGRHYALKP DAY Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
HOXC9 belongs to the homeobox family. The homeobox family is a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12.
Especificaciones
Especificaciones
| Antígeno | HOXC9 |
| Aplicaciones | ChIP Assay, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.4 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | HOXC9 |
| N.º de referencia del gen | P31274 |
| Alias de gen | homeo box C9; homeobox C9; homeobox protein Hox-3.2; Homeobox protein Hox-3B; Homeobox protein Hox-C9; HOX3; Hox-3.2; HOX3B; Hoxc8; Hoxc9; HOXC-9 |
| Símbolos de los genes | HOXC9 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?