missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXC8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5116256
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HOXC8 Polyclonal Antibody for Western Blot
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene may play a role in the regulation of cartilage differentiation. It could also be involved in chondrodysplasias or other cartilage disorders.
Especificaciones
| HOXC8 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P09025 | |
| HOXC8 | |
| KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of mouse Hoxc8. | |
| 400 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| HOXC8 | |
| D130011F21Rik; homeo box 3A; homeo box C8; homeobox C8; Homeobox gene C8; homeobox protein Hox-3.1; Homeobox protein Hox-3A; homeobox protein Hox-C8; Homeobox protein M31; homeobox protein R4; HOX3; Hox-3.1; Hox-3.1, mouse, homolog of; HOX3A; Hox3r4; HOXC8; Hoxc-8 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 15426 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?