missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: EVTFREYAIE PATKWHPRGN LAHCYSAEEL VHRDCLQAPS AAGVPGDVLA KSSANVYHHP TPAVSSNFY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
HOX11, also called homeobox11 or Tcl-3 is located at the 10q24 T-Cell translocation breakpoint region. It encodes a homeobox-domain containing protein, HOX11, containing a glycine and proline-rich amino terminus. HOX11 is required for maintenance of the developing spleen and cell survival. The HOX11protein binds to a specific DNA sequence and localizes to the cell nucleus. It transactivates transcription of a reporter gene linked to a cis-regulatory element, acting as a positive transcription activator. The HOX11 gene is highly expressed in T-ALLs as a result of a t (7:10) (q35;q24) or t (10:14) (q24;qll) translocation. The homeobox gene is deregulated upon translocation into the T-cell receptor locus and activates a specific subset of tumor-associated target genes. HOX 11 gene translocation has been attributed to T-cell leukemia and lymphoid neoplasias.
Especificaciones
Especificaciones
| Antígeno | HOXA11 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | HOXA11 |
| N.º de referencia del gen | P31270 |
| Alias de gen | homeo box 1I; homeo box A11; homeobox A11; homeobox protein Hox-1.9; Homeobox protein Hox-1I; homeobox protein HOXA11; homeobox protein Hox-A11; HOX1; Hox-1.9; HOX11; HOX1I; Hoxa11; Hoxa-11; NEWGENE_1564605; RUSAT1 |
| Símbolos de los genes | HOXA11 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
