Learn More
HNRNPA1 Mouse, anti-Human, Unconjugated, Clone: 1A5A4, Proteintech
Código de producto. 17271506 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17271506 20 μL 20 microlitros
17261506 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17271506 Proveedor Proteintech N.º de proveedor 678441IG20UL
Mouse Monoclonal Antibody
In eukaryotic cells, nascent RNA polymerase II transcripts are associated in the nucleus with specific proteins to form ribonucleoprotein complexes called HNRP or 40S. Protein moiety of the HNRP particle has 6 major components called core proteins, and HNRPA1 is one of them. hnRNP A1 is a multifunctional RNA binding protein which is involved in pre-mRNA splicing, export of mature transcripts from the nucleus, mRNA turnover, and internal ribosome entry site (IRES)-mediated translation. HNRNPA1 has three isoforms with MW 30 kDa, 34 kDa and 39 kDa.
Especificaciones
Antígeno HNRNPA1
Aplicaciones Western Blot, ELISA
Clasificación Monoclonal
Clon 1A5A4
Concentración 500 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 3178
Especies diana Human, Mouse