missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP A1 Polyclonal Antibody, Invitrogen™
Descripción
Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with mouse, rat and macaque monkey based on sequence homology. Store antibody at 4oC for 1-2 weeks. For long-term storage, store at -20oC.
HnRNP A1 belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre mRNAs in the nucleus and appear to influence pre mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. hnRNP A1 has two repeats of quasi RRM domains that bind to RNAs. It is one of the most abundant core proteins of hnRNP complexes and it is localized to the nucleoplasm. This protein, along with other hnRNP proteins, is exported from the nucleus, probably bound to mRNA, and is immediately re imported.
Especificaciones
Especificaciones
| Antígeno | hnRNP A1 |
| Aplicaciones | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.9 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA and 0.02% sodium azide; pH 7.4 |
| génica | Hnrnpa1 |
| N.º de referencia del gen | P09651 |
| Alias de gen | ALS19; ALS20; Fli-2; Hdp; HDP-1; Helix-destabilizing protein; heterogeneous nuclear ribonucleoprotein A1; Heterogeneous nuclear ribonucleoprotein A1, N-terminally processed; heterogeneous nuclear ribonucleoprotein A1B protein; heterogeneous nuclear ribonucleoprotein B2 protein; heterogeneous nuclear ribonucleoprotein core protein A1; hnRNP A1; hnRNP A1-like 3; hnRNP core protein A1; hnRNP core protein A1-like 3; hnRNP I; Hnrnpa1; hnRNP-A1; HNRNPI; HNRNP-I; Hnrpa1; HNRPA1L3; HNRPA1MGC102835; HNRPI; I79_022927; IBMPFD3; MGC102835; MGC128297 protein; nuclear ribonucleoprotein particle A1 protein; pPTB; PTB; PTB-1; PTB2; PTB3; PTB4; PTB-T; putative heterogeneous nuclear ribonucleoprotein A1-like 3; Putative heterogeneous nuclear ribonucleoprotein A1-like protein 3; ROA1; RP11-78J21.1; single-strand DNA-binding protein UP1; single-strand RNA-binding protein; Single-strand-binding protein; Tis; Topoisomerase-inhibitor suppressed; unwinding protein 1; UP 1; UP1 |
| Símbolos de los genes | Hnrnpa1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?