missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF4A, Mouse anti-Human, Clone: 4E2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00003172-M04.50ug
This item is not returnable.
View return policy
Description
Sequence: NDRQYDSRGRFGELLLLLPTLQSITWQMIEQIQFIKLFGMAKIDNLLQEMLLGGSPSDAPHAHHPLHPHLMQEHMGTNVIVANTMPTHLSNGQMCEWPRPSpecifications
| HNF4A | |
| Monoclonal | |
| Unconjugated | |
| hepatocyte nuclear factor 4, alpha | |
| FLJ39654/HNF4/HNF4a7/HNF4a8/HNF4a9/MODY/MODY1/NR2A1/NR2A21/TCF/TCF14 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, KnockDown, Western Blot | |
| 4E2 | |
| In 1x PBS, pH 7.4 | |
| NM_000457 | |
| HNF4A | |
| HNF4A (NP_000448, 324 a.a. ∼ 423 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 3172 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction